siRNA Details
Gene Symbol: Uvssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081101.2 | NP_001074570.1 | 3, 4 | 746 | ||
GenBank mRNA | AK016722.1 | - | 2, 3 | 407 | ||
AK028507.1 | - | 2, 3 | 1115 | |||
AK040506.1 | - | 3, 4 | 750 | |||
AK129384.1 | - | 3, 4 | 432 | |||
AK156117.1 | - | 2, 3 | 905 | |||
BC137803.1 | - | 3, 4 | 670 | 8860822 | ||
BC144920.1 | - | 3, 4 | 585 | 9053468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
