siRNA Details
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039081.2 | NP_001034170.1 | 12 | 1197 | ||
NM_001286221.1 | NP_001273150.1 | 14 | 1328 | |||
NM_001286222.1 | NP_001273151.1 | 14 | 1382 | |||
NM_001286223.1 | NP_001273152.1 | 12 | 1178 | |||
NM_027696.3 | NP_081972.2 | 14 | 1385 | |||
GenBank mRNA | AK016915.1 | - | 6 | 627 | ||
AK043136.1 | - | 1 | 1116 | |||
AK129405.1 | - | 14 | 1305 | |||
AK136467.1 | - | 2 | 41 | |||
AY769435.1 | - | 13 | 1177 | |||
AY769436.1 | - | 12 | 1100 | |||
AY769437.1 | - | 12 | 1150 | |||
AY769438.1 | - | 14 | 1250 | |||
BC108273.1 | - | 12 | 1164 | 30080243 | ||
BC130227.1 | - | 12 | 1149 | 40141863 | ||
CN701986.1 | - | 6 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
