siRNA Details
Gene Symbol: Dydc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027717.1 | NP_081993.1 | 2 | 196 | ||
GenBank mRNA | AK016963.1 | - | 2 | 331 | ||
BC113750.1 | - | 2 | 196 | 40049377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
