siRNA Details
Gene Symbol: Sdhaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077713.1 | NP_001071181.1 | 2 | 329 | ||
GenBank mRNA | AK002229.1 | - | 2 | 311 | ||
AK004417.1 | - | 2 | 259 | |||
AK028031.1 | - | 2 | 300 | |||
AK046436.1 | - | 344 | ||||
BC118927.1 | - | 2 | 329 | 40090186 | ||
BC118928.1 | - | 2 | 336 | 40090189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
