siRNA Details
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023596.3 | NP_076085.1 | 5 | 792 | ||
GenBank mRNA | AF326986.1 | - | 5 | 803 | ||
AK017065.1 | - | 2 | 687 | |||
AK034133.1 | - | 5 | 788 | |||
AK051317.1 | - | 5 | 817 | |||
AK210732.1 | - | 1 | 66 | |||
BC137864.1 | - | 5 | 775 | 8860883 | ||
BC144983.1 | - | 5 | 775 | |||
CV557129.1 | - | 4 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
