siRNA Details
Gene Symbol: Treml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289451.1 | NP_001276380.1 | 3 | 450 | ||
NM_001289457.1 | NP_001276386.1 | 3 | 450 | |||
NM_027763.2 | NP_082039.1 | 3 | 450 | |||
GenBank mRNA | AK017256.1 | - | 3 | 451 | ||
AK079428.1 | - | 3 | 451 | |||
AY078502.1 | - | 3 | 450 | |||
AY286071.1 | - | 3 | 450 | |||
BC138399.1 | - | 3 | 449 | 8861419 | ||
BC145263.1 | - | 3 | 449 | 9053816 | ||
DQ087181.1 | - | 3 | 218 | |||
DQ087182.1 | - | 2 | 599 | |||
DQ087183.1 | - | 3 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
