siRNA Details
Gene Symbol: Col24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027770.2 | NP_082046.2 | 38, 39 | 4055 | ||
GenBank mRNA | BC139134.1 | - | 38, 39 | 3612 | 8862156 | |
BC145544.1 | - | 38, 39 | 3604 | |||
DQ157748.1 | - | 38, 39 | 4055 |
