siRNA Details
Gene Symbol: Foxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183186.2 | NP_899009.2 | 3 | 652 | ||
GenBank mRNA | AK017346.1 | - | 2 | 455 | ||
AK046748.1 | - | 3 | 927 | |||
BC099971.1 | - | 3 | 730 | 30008715 | ||
BY717160.1 | - | 2 | 450 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108047, XM_006240429, XM_008764802, XM_008764801, XM_008764803, and others.
Gene Ontology Categories:
