siRNA Details
Gene Symbol: Foxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183186.2 | NP_899009.2 | 3 | 646 | ||
GenBank mRNA | AK017346.1 | - | 2 | 449 | ||
AK046748.1 | - | 3 | 921 | |||
BC099971.1 | - | 3 | 724 | 30008715 | ||
BY717160.1 | - | 2 | 444 |
