siRNA Details
Gene Symbol: Kctd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017384.1 | - | 3 | 318 | ||
AK082924.1 | - | 3 | 317 | |||
AK159067.1 | - | 3 | 1730 | |||
BB625950.1 | - | 3 | 207 | |||
BC096370.1 | - | 3 | 157 | 3597940 | ||
BC131971.1 | - | 2 | 127 | 40130248 | ||
BC131975.1 | - | 2 | 127 | 40130252 | ||
CX239507.1 | - | 3 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
