siRNA Details
Gene Symbol: 5430416O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017317.1 | - | 2 | 741 | ||
AK030334.1 | - | 2 | 759 | |||
BC064043.1 | - | 2 | 726 | |||
BC147344.1 | - | 1 | 290 | 9056418 | ||
BC147345.1 | - | 1 | 290 | 9056411 |
