siRNA Details
Gene Symbol: Obox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027802.2 | NP_082078.1 | 2 | 145 | ||
GenBank mRNA | AK018362.1 | - | 2 | 142 | ||
AK136061.1 | - | 2 | 135 | |||
BC141323.1 | - | 105 | 9055935 | |||
BC141324.1 | - | 105 | 9055936 |
Gene Symbol: Obox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145708.2 | NP_663754.2 | 2 | 242 | ||
GenBank mRNA | BC094912.1 | - | 2 | 122 | ||
CA556780.1 | - | 2 | 142 | |||
DV663188.1 | - | 2 | 241 |
