siRNA Details
Gene Symbol: Zfp935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136496.1 | NP_001129968.1 | 3 | 471 | ||
NM_178875.4 | NP_849206.2 | 4 | 532 | |||
GenBank mRNA | AK018441.1 | - | 3 | 462 | ||
AY189975.1 | - | 3 | 448 | |||
BC012524.1 | - | 3 | 447 | 4008347 | ||
BC037448.1 | - | 3 | 476 | |||
BC047396.1 | - | 4 | 509 | 3979342 | ||
BC087936.1 | - | 4 | 532 | |||
BC113193.1 | - | 4 | 528 | 40062446 | ||
BC115966.1 | - | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
