siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 27 | 2755 | ||
GenBank mRNA | AK043149.1 | - | 1 | 527 | ||
AK050302.1 | - | 27 | 2287 | |||
BC007154.1 | - | 677 | ||||
BC025808.1 | - | 12 | 942 | |||
BC038055.1 | - | 22 | ||||
BC043674.1 | - | 20 | 1732 | 5044565 | ||
BC054720.1 | - | 27 | 2273 | 5689248 |
