siRNA Details
Gene Symbol: Cyp2u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027816.3 | NP_082092.2 | 2 | 922 | ||
GenBank mRNA | AK018458.1 | - | 2 | 898 | ||
AK041477.1 | - | 2, 3 | 818 | |||
AK142740.1 | - | 2 | 863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
