siRNA Details
Gene Symbol: Kazn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109684.1 | NP_001103154.1 | 5 | 1157 | ||
NM_001109685.1 | NP_001103155.1 | 6 | 1175 | |||
NM_144531.3 | NP_653114.3 | 5 | 1157 | |||
GenBank mRNA | AK018497.1 | - | 6 | 1155 | ||
AK032460.1 | - | 3 | 396 | |||
AK052809.1 | - | 6 | 1176 | |||
AK173090.1 | - | 5 | 1157 | |||
BC022941.1 | - | 5 | 1161 | |||
BC100387.1 | - | 3 | 382 | |||
CD349067.1 | - | 5 | 732 |

More Information
Other Species Targeted

Gene Ontology Categories:
