siRNA Details
Gene Symbol: Kazn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144531.3 | NP_653114.3 | 12 | 2066 | ||
GenBank mRNA | AK032460.1 | - | 10 | 1305 | ||
AK173090.1 | - | 12 | 2066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
