siRNA Details
Gene Symbol: Fam217b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081289.1 | NP_001074758.1 | 4 | 903 | ||
GenBank mRNA | AK155442.1 | - | 4 | 905 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam217b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081289.1 | NP_001074758.1 | 4 | 903 | ||
GenBank mRNA | AK155442.1 | - | 4 | 905 |