siRNA Details
Gene Symbol: 9130008F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027834.3 | NP_082110.1 | 1 | 349 | ||
GenBank mRNA | AK018601.1 | - | 1 | 350 | ||
AK033670.1 | - | 1 | 345 | |||
BC132063.1 | - | 1 | 345 | 40130340 | ||
BC132065.1 | - | 1 | 345 | 40130342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
