siRNA Details
Gene Symbol: Senp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172068.1 | NP_001165539.1 | 4 | 409 | ||
NM_001172069.1 | NP_001165540.1 | 3 | 318 | |||
NM_001172070.1 | NP_001165541.1 | 3 | 299 | |||
NM_001172071.1 | NP_001165542.1 | 3 | 316 | |||
NM_027838.3 | NP_082114.1 | 2 | 206 | |||
GenBank mRNA | AK018606.1 | - | 3 | 297 | ||
AK029856.1 | - | 2 | 196 | |||
AK049120.1 | - | 4 | 399 | |||
AK136214.1 | - | 3 | 289 | |||
BC037443.1 | - | 2 | 178 | 3599077 | ||
CO431667.1 | - | 3 | 283 | |||
DV645349.1 | - | 2 | 206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
