siRNA Details
Gene Symbol: 9130011E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198296.2 | NP_938038.2 | 7 | 644 | ||
GenBank mRNA | AK159765.1 | - | 7 | 645 | ||
AK188005.1 | - | 3 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
