siRNA Details
Gene Symbol: Arl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027843.1 | NP_082119.1 | 1 | 205 | ||
GenBank mRNA | AK018621.1 | - | 1 | 206 | ||
BC104368.1 | - | 1 | 166 | 40049874 | ||
BC104369.1 | - | 1 | 166 | 40049875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
