siRNA Details
Gene Symbol: Rnf215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027859.2 | NP_082135.2 | 9 | 1432 | ||
GenBank mRNA | AK002414.1 | - | 9 | 1390 | ||
AK154140.1 | - | 7 | 1335 | |||
BC103627.1 | - | 9 | 1356 | 6486672 | ||
BM948157.1 | - | 1 | 21 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
