siRNA Details
Gene Symbol: 0610010F05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027860.2 | NP_082136.2 | 6 | 915 | ||
GenBank mRNA | AK053838.1 | - | 6 | 912 | ||
AK129454.1 | - | 6 | 882 | |||
BC055049.1 | - | 5 | 788 | |||
BC065122.1 | - | 7 | 922 | |||
BC072622.1 | - | 6 | 905 | 6821718 | ||
BC079876.1 | - | 6 | 901 | 6848269 |
