siRNA Details
Gene Symbol: Atp5h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027862.1 | NP_082138.1 | 6 | 410 | ||
GenBank mRNA | AF354051.1 | - | 5 | 403 | ||
AK002376.1 | - | 6 | 410 | |||
AK088617.1 | - | 5 | 405 | |||
BC016547.1 | - | 6 | 408 | 2631309 | ||
BC081431.1 | - | 6 | 409 | 6408035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
