siRNA Details
Gene Symbol: Esm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023612.3 | NP_076101.1 | 1 | 276 | ||
GenBank mRNA | AJ249354.1 | - | 1 | 139 | ||
AK002913.1 | - | 1 | 170 | |||
BC020038.1 | - | 1 | 149 | 4018658 | ||
BC051919.1 | - | 1 | 276 | 6403364 | ||
BC147648.1 | - | 1 | 144 | 9056638 | ||
BC147650.1 | - | 1 | 144 | 9056640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
