siRNA Details
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027866.1 | NP_082142.1 | 7 | 691 | ||
GenBank mRNA | AK003121.1 | - | 7 | 691 | ||
BC104380.1 | - | 7 | 662 | 40050136 | ||
BC104381.1 | - | 7 | 680 | 40050143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
