siRNA Details
Gene Symbol: Dram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027878.2 | NP_082154.2 | 5 | 691 | ||
GenBank mRNA | AK004552.1 | - | 5 | 691 | ||
AK151184.1 | - | 5 | 624 | |||
BC021433.1 | - | 5 | 657 | 5030012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
