siRNA Details
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027879.2 | NP_082155.1 | 3 | 438 | ||
GenBank mRNA | AK004569.1 | - | 3 | 405 | ||
AK040680.1 | - | 3 | 388 | |||
AK084140.1 | - | 3 | 436 | |||
AK135411.1 | - | 3 | 405 | |||
BC061220.1 | - | 3 | 413 | |||
BC147503.1 | - | 3 | 438 | 9056386 | ||
BC158078.1 | - | 3 | 438 | 9088170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
