siRNA Details
Gene Symbol: Stk11ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027886.3 | NP_082162.3 | 18 | 2225 | ||
GenBank mRNA | AK004757.2 | - | 18 | 2178 | ||
AK156122.1 | - | 17 | 2313 | |||
AK165911.1 | - | 18 | 2217 | |||
AK171989.1 | - | 17 | 2087 | |||
AK195907.1 | - | 3 | 213 | |||
AK213443.1 | - | 2 | 131 | |||
BC035530.1 | - | 4 | 1265 | |||
BC042403.1 | - | 16 | 2262 | |||
BC061635.1 | - | 18 | 2208 | 6530178 |
