siRNA Details
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027891.4 | NP_082167.2 | 8 | 1117 | ||
GenBank mRNA | AK004717.1 | - | 6 | 2079 | ||
AK051910.1 | - | 8 | 1065 | |||
AK084948.1 | - | 8 | 1117 | |||
AK153976.1 | - | 8 | 1088 | |||
AK154352.1 | - | 8 | 1059 | |||
AK167020.1 | - | 8 | 1066 | |||
AK172708.1 | - | 8 | 1085 | |||
BC048397.1 | - | 8 | 1096 | 6396815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
