siRNA Details
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 4, 5 | 506 | ||
GenBank mRNA | AK004875.2 | - | 4, 5 | 462 | ||
AK016520.1 | - | 4, 5 | 476 | |||
AK028589.1 | - | 4, 5 | 475 | |||
AK144693.1 | - | 4, 5 | 479 | |||
AK160597.1 | - | 4, 5 | 491 | |||
AK220287.1 | - | 4, 5 | 481 | |||
BC026779.1 | - | 4, 5 | 407 | 4018298 | ||
BC027428.1 | - | 4, 5 | 469 | 3983461 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108417, XM_006254072, XM_006254074, XM_006254073.
Gene Ontology Categories:
