siRNA Details
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 3 | 408 | ||
GenBank mRNA | AK016520.1 | - | 3 | 378 | ||
AK028589.1 | - | 3 | 377 | |||
AK144693.1 | - | 3 | 381 | |||
AK160597.1 | - | 3 | 393 | |||
AK220287.1 | - | 3 | 383 | |||
BC026779.1 | - | 3 | 309 | 4018298 | ||
BC027428.1 | - | 3 | 371 | 3983461 |
