siRNA Details
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 21 | 2341 | ||
GenBank mRNA | AK004875.2 | - | 21 | 2297 | ||
AK016520.1 | - | 20 | 2194 | |||
AK028589.1 | - | 21 | 2310 | |||
AK144693.1 | - | 21 | 2315 | |||
AK160597.1 | - | 21 | 2326 | |||
AK220287.1 | - | 21 | 2316 | |||
BC025902.1 | - | 13 | 1407 | |||
BC026779.1 | - | 21 | 2242 | 4018298 | ||
BC027428.1 | - | 21 | 2304 | 3983461 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108417, XM_006254072, XM_006254074, XM_006254073.
Gene Ontology Categories:
