siRNA Details
Gene Symbol: R3hdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168292.1 | NP_001161764.1 | 7 | 674 | ||
NM_001168293.1 | NP_001161765.1 | 6 | 989 | |||
NM_027900.4 | NP_082176.4 | 6 | 989 | |||
GenBank mRNA | AK043545.1 | - | 6 | 990 | ||
AK122416.1 | - | 6 | 532 | |||
BC043083.1 | - | 4 | 286 | |||
BC053099.1 | - | 2 | 93 | |||
BC064442.1 | - | 2 | 61 | |||
BC117927.2 | - | 6 | 732 | 40097626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
