siRNA Details
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163729.2 | NP_001157201.2 | 8 | 1184 | ||
NM_027910.3 | NP_082186.2 | 9 | 1337 | |||
GenBank mRNA | AB053465.1 | - | 9 | 1208 | ||
AJ278127.1 | - | 9 | 1204 | |||
AK004964.1 | - | 9 | 1314 | |||
AK079082.1 | - | 9 | 1217 | |||
AK141673.1 | - | 8 | 1169 | |||
AK170623.1 | - | 9 | 1209 | |||
BC018154.1 | - | 9 | 1306 | 3983436 |
