siRNA Details
Gene Symbol: Ranbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252466.1 | NP_001239395.1 | 8 | 783 | ||
NM_001252467.1 | NP_001239396.1 | 5 | 423 | |||
NM_027933.3 | NP_082209.1 | 7 | 582 | |||
GenBank mRNA | AK040481.1 | - | 9 | 766 | ||
AK133949.1 | - | 7 | 429 | |||
AK154201.1 | - | 6 | 1986 | |||
AK155301.1 | - | 7 | 451 | |||
AK167731.1 | - | 7 | 432 | |||
BC066005.1 | - | 7 | 423 | |||
BC145892.1 | - | 7 | 446 | 40131195 | ||
BC145894.1 | - | 7 | 446 | 40131197 | ||
BM950080.1 | - | 7 | 517 | |||
CA318306.1 | - | 8 | 618 | |||
CN717165.1 | - | 5 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
