siRNA Details
Gene Symbol: Tmem50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027935.2 | NP_082211.1 | 3 | 191 | ||
GenBank mRNA | AK014282.1 | - | 3 | 261 | ||
AK147069.1 | - | 3 | 227 | |||
AK152630.1 | - | 3 | 275 | |||
AK152654.1 | - | 3 | 246 | |||
AK157214.1 | - | 3 | 264 | |||
AK164989.1 | - | 3 | 257 | |||
AK165636.1 | - | 3 | 240 | |||
BC094256.1 | - | 3 | 191 | 6820666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
