siRNA Details
Gene Symbol: Dcaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027946.3 | NP_082222.1 | 3 | 643 | ||
GenBank mRNA | AK005902.1 | - | 3 | 637 | ||
AK047435.1 | - | 3 | 645 | |||
AK080823.1 | - | 3 | 617 | |||
AK154757.1 | - | 3 | 634 | |||
AK165674.1 | - | 3 | 634 | |||
BB646282.1 | - | 3 | 644 | |||
BC048165.1 | - | 3 | 559 | 6409531 | ||
BC048722.1 | - | 3 | 562 | 6504571 |
