siRNA Details
Gene Symbol: Lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133737.2 | NP_598498.1 | 6 | 1533 | ||
GenBank mRNA | AB035967.1 | - | 6 | 1352 | ||
AK005632.1 | - | 6 | 902 | |||
BC016072.1 | - | 6 | 1387 | 4482042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
