siRNA Details
Gene Symbol: Lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133737.2 | NP_598498.1 | 2 | 825 | ||
GenBank mRNA | AB035967.1 | - | 2 | 644 | ||
AK005632.1 | - | 2 | 194 | |||
BC016072.1 | - | 2 | 679 | 4482042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
