siRNA Details
Gene Symbol: Lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133737.2 | NP_598498.1 | 6 | 1457 | ||
GenBank mRNA | AB035967.1 | - | 6 | 1276 | ||
AK005632.1 | - | 6 | 826 | |||
BC016072.1 | - | 6 | 1311 | 4482042 |
