siRNA Details
Gene Symbol: Shcbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033162.2 | NP_001028334.1 | 6 | 1065 | ||
GenBank mRNA | AK005886.1 | - | 6 | 1062 | ||
AK161274.1 | - | 6 | 1066 | |||
AK171525.1 | - | 6 | 1103 | |||
BC113157.1 | - | 6 | 1112 | 40058375 |

More Information
Other Species Targeted

Gene Ontology Categories:
