siRNA Details
Gene Symbol: Osgin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027950.1 | NP_082226.1 | 6 | 739 | ||
GenBank mRNA | AB221676.1 | - | 5 | 566 | ||
AK005892.1 | - | 4 | 502 | |||
AK156249.1 | - | 1 | 1228 | |||
BC006032.1 | - | 5 | 1080 | |||
BC022135.1 | - | 6 | 739 | 5099665 | ||
BC110674.1 | - | 6 | 724 | |||
BC132440.1 | - | 6 | 708 | 40130717 | ||
BC138192.1 | - | 6 | 708 | 8861211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
