siRNA Details
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168244.1 | NP_001161716.1 | 3 | 816 | ||
NM_001168246.1 | NP_001161718.1 | 4 | 515 | |||
NM_027954.3 | NP_082230.2 | 4 | 497 | |||
GenBank mRNA | AK005954.1 | - | 4 | 498 | ||
AK082833.1 | - | 3 | 383 | |||
AK167332.1 | - | 3 | 815 | |||
BC094628.1 | - | 4 | 392 | 3374002 | ||
BC132534.1 | - | 4 | 447 | 40130811 | ||
BY083329.1 | - | 3 | 229 |

More Information
Other Species Targeted

Gene Ontology Categories:
