siRNA Details
Gene Symbol: Fam178b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126046.1 | NP_001119518.1 | 12, 13 | 1720 | ||
NM_027957.1 | NP_082233.1 | 3, 4 | 343 | |||
GenBank mRNA | AK006300.1 | - | 3, 4 | 344 | ||
BC114576.1 | - | 3, 4 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
