siRNA Details
Gene Symbol: Gpr160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134385.2 | NP_001127857.1 | 4 | 1716 | ||
NM_001134386.2 | NP_001127858.1 | 3 | 1199 | |||
NM_001286994.1 | NP_001273923.1 | 4 | 1725 | |||
NM_001286995.1 | NP_001273924.1 | 3 | 1428 | |||
NM_001286996.1 | NP_001273925.1 | 4 | 1299 | |||
NM_027965.2 | NP_082241.1 | 4 | 1528 | |||
GenBank mRNA | AK006330.1 | - | 4 | 1488 | ||
AK154620.1 | - | 3 | 1201 | |||
AK154784.1 | - | 4 | 1683 | |||
BC116649.1 | - | 4 | 1651 | 40088354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
