siRNA Details
Gene Symbol: 1700019O17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027966.1 | NP_082242.1 | 1 | 670 | ||
GenBank mRNA | AK006136.1 | - | 1 | 670 | ||
BC100497.1 | - | 1 | 657 | 30916756 | ||
BC145753.1 | - | 2 | 579 | 40131056 |
