siRNA Details
Gene Symbol: Fbxo30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168297.1 | NP_001161769.1 | 2 | 1295 | ||
NM_027968.3 | NP_082244.2 | 2 | 1329 | |||
GenBank mRNA | AK006369.1 | - | 2 | 1296 | ||
AK083479.1 | - | 2 | 1286 | |||
AK135274.1 | - | 2 | 1288 | |||
BC037691.1 | - | 1 | 208 | 3601336 |
