siRNA Details
Gene Symbol: 2310007B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159940.1 | NP_001153412.1 | 4 | 1135 | ||
NM_172411.4 | NP_765999.3 | 4 | 1126 | |||
GenBank mRNA | AK009193.1 | - | 4 | 1135 | ||
AK081300.1 | - | 3 | 1014 | |||
BC089525.1 | - | 4 | 1126 | 30315016 |

More Information
Other Species Targeted

Gene Ontology Categories:
